missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PF 06424439

Click to view available options
:
5mg
Description
PF 06424439 is a potent and selective DGAT2 inhibitor (IC50 = 14 nM). Exhibits no significant activity at MGAT1-3 or DGAT1. Reduces triglyderide synthesis in human hepatocytes in vitro. Reduces plasma triglyceride and cholesterol levels in a rat dyslipidemia model. Orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | [(3R)-1-[2-[1-(4-Chloro-1H-pyrazol-1-yl)cyclopropyl]-3H-imidazo[4,5-b]pyridin-5-yl]-3-piperidinyl]-1-pyrrolidinylmethanone methanesulfonate |
| CAS | 1469284-79-4 |
| Assay Percent Range | >98% |
| Quantity | 5 mg |
| Molecular Formula | C22H26ClN7O.CH4O3S |
| Formula Weight | Observed MW: 536.05 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction