missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PF 06409577
GREENER_CHOICE

Product Code. p-7121041
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18756559

Brand: Tocris Bioscience™ 6114/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective allosteric activator of AMPK

PF 06409577 is a potent and selective allosteric activator of AMPK (EC50 values are 7 and >40000 nM for AMPKα1β1γ1 and AMPKα1β2γ1, respectively). Exhibits minimal off-target effects on a panel of receptors, ion channels, PDEs and kinases. Improves kidney function and reduces proteinuria in a preclinical model of diabetic nephropathy. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material PF 06409577
CAS 1467057-23-3
Synonym 6-Chloro-5-[4-(1-hydroxycyclobutyl)phenyl]-1H-indole-3-carboxylic acid
Quantity 50 mg
Solubility Soluble to 100mM in DMSO and to 20mM in ethanol
Molecular Formula C19H16ClNO3
Formula Weight 341.79
Physical Form Solid
Color Gray
Product Type Bioactive Small Molecule
Purity 99%
Show More Show Less

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.