missing translation for 'onlineSavingsMsg'
Learn More

PF-06260933, MedChemExpress

Product Code. 16443767
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16443767

Brand: Medchem Express HY19562

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PF-06260933 is an orally active and highly selective inhibitor of MAP4K4 with IC50s of 3.7 and 160 nM for kinase and cell, respectively.

PF-06260933 is an orally active and highly selective inhibitor of MAP4K4 with IC50s of 3.7 and 160 nM for kinase and cell, respectively.

Specifications

Chemical Name or Material PF-06260933
CAS 1811510-56-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C16H13ClN4
Quantity 50 mg
Solubility Information DMSO : 30 mg/mL (101.10 mM; Need ultrasonic and warming)
SMILES NC1=C(C2=CC=C(Cl)C=C2)C=C(C(C=N3)=CC=C3N)C=N1
Molecular Weight (g/mol) 296.75
Formula Weight 296.75
Percent Purity 98.41%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.