missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PF 05175157

Product Code. 18754739
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18754739 10 mg 10mg
18789228 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18754739 Supplier Tocris Bioscience™ Supplier No. 5790/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Acetyl-CoA carboxylase (ACC) 1 and 2 inhibitor

PF 05175157 is an potent acetyl-CoA carboxylase (ACC) 2 and 1 inhibitor (IC50 values are 45 and 98 nM, respectively). Reduces hepatic and skeletal muscle malonyl-CoA levels in rats. Also inhibits hepatic de novo lipogenesis and reduces whole body respiratory exchange ratio in rats. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,4-Dihydro-1′-[2-methyl-1H-benzimidazol-6-yl)carbonyl]-1-(1-methylethyl)-spiro[5H-indazole-5,4′-piperidin]-7(6H)-one
CAS 1301214-47-0
Quantity 10 mg
Target Additional Lipid Metabolism Compounds
Molecular Formula C23H27N5O2
Formula Weight 405.49
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.