missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PF 05089771, MedChemExpress
Click to view available options
Quantity:
5 mg
100 mg
Unit Size:
100mg
5mg
Description
PF 05089771 is a potent, orally active and selective arylsulfonamide Nav1.7 inhibitor, with IC50 values of 11 nM, 12 nM, 13 nM, 171 nM and 8 nM for hNav1.7, cynNav1.7, dogNav1.7,
Specifications
Specifications
| Chemical Name or Material | PF 05089771 |
| CAS | 1235403-62-9 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C18H12Cl2FN5O3S2 |
| Quantity | 5 mg |
| Solubility Information | DMSO : ≥ 34 mg/mL (67.95 mM) |
| SMILES | O=S(C1=CC(Cl)=C(OC2=CC=C(Cl)C=C2C3=CNN=C3N)C=C1F)(NC4=CSC=N4)=O |
| Molecular Weight (g/mol) | 500.35 |
| Formula Weight | 500.35 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction