missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PF 05089771

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
PF 05089771 is a potent and selective Nav1.7 channel blocker (IC50 = 8, 11 and 171 nM for mouse, human and rat Nav1.7, respectively). Exhibits selectivity for Nav1.7 over other Nav1 channels (IC50 values are 0.11, 0.16, 0.85, 10, 11 and 25 μM for Nav1.2, Nav1.6, Nav1.1, Nav1.4, Nav1.3 and Nav1.5, respectively). Also exhibits selectivity over a panel of 81 other ion channels, receptors, enzymes and transporters. Blocks spontaneous firing of inherited erythromelalgia (IEM)-derived iPSC sensory neurons in vitro.
Specifications
Specifications
| Chemical Name or Material | 4-[2-(3-Amino-1H-pyrazol-4-yl)-4-chlorophenoxy]-5-chloro-2-fluoro-N-4-thiazolylbenzenesulfonamide tosylate |
| CAS | 1430806-04-4 |
| Quantity | 10 mg |
| Target | Voltage-gated Sodium Channel Blockers |
| Molecular Formula | C18H12Cl2FN5O3S2.C7H8O3S |
| Formula Weight | 672.56 |
| Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction