missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PF 05089771
GREENER_CHOICE

Product Code. 18709248
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18709248

Brand: Tocris Bioscience™ 5931/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective Nav1.7 channel blocker

PF 05089771 is a potent and selective Nav1.7 channel blocker (IC50 = 8, 11 and 171 nM for mouse, human and rat Nav1.7, respectively). Exhibits selectivity for Nav1.7 over other Nav1 channels (IC50 values are 0.11, 0.16, 0.85, 10, 11 and 25 μM for Nav1.2, Nav1.6, Nav1.1, Nav1.4, Nav1.3 and Nav1.5, respectively). Also exhibits selectivity over a panel of 81 other ion channels, receptors, enzymes and transporters. Blocks spontaneous firing of inherited erythromelalgia (IEM)-derived iPSC sensory neurons in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-[2-(3-Amino-1H-pyrazol-4-yl)-4-chlorophenoxy]-5-chloro-2-fluoro-N-4-thiazolylbenzenesulfonamide tosylate
CAS 1430806-04-4
Quantity 10 mg
Target Voltage-gated Sodium Channel Blockers
Molecular Formula C18H12Cl2FN5O3S2.C7H8O3S
Formula Weight 672.56
Purity >98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.