missing translation for 'onlineSavingsMsg'
Learn More

PF-05085727, MedChemExpress

Product Code. p-200097398
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16483698

Brand: Medchem Express HY102050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PF-05085727 is a potent, selective and brain penetrant inhibitor of cGMP-dependent PDE2A (IC50=2 nM). PF-05085727 inhibits PDE2A >4,000-fold selectivity over PDE1 and PDE3-11.

PF-05085727 is a potent, selective and brain penetrant inhibitor of cGMP-dependent PDE2A (IC50=2 nM). PF-05085727 inhibits PDE2A >4,000-fold selectivity over PDE1 and PDE3-11.

Specifications

Chemical Name or Material PF-05085727
CAS 1415637-72-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C20H18F3N7
Quantity 10 mg
Solubility Information DMSO : 62.5 mg/mL (151.19 mM; Need ultrasonic)
SMILES CN1C2=NC=NC(N3CCC3)=C2C(C4=C(C5=CC=C(C(F)(F)F)C=C5)N(C)N=C4)=N1
Molecular Weight (g/mol) 413.4
Formula Weight 413.4
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.