missing translation for 'onlineSavingsMsg'
Learn More

PF-05085727, MedChemExpress

Product Code. 16483698
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16483698 10 mg 10mg
16476437 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16483698 Supplier Medchem Express Supplier No. HY102050
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress PF-05085727 is a potent, selective and brain penetrant inhibitor of cGMP-dependent PDE2A (IC50=2 nM). PF-05085727 inhibits PDE2A >4,000-fold selectivity over PDE1 and PDE3-11.

PF-05085727 is a potent, selective and brain penetrant inhibitor of cGMP-dependent PDE2A (IC50=2 nM). PF-05085727 inhibits PDE2A >4,000-fold selectivity over PDE1 and PDE3-11.

Specifications

Chemical Name or Material PF-05085727
CAS 1415637-72-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C20H18F3N7
Quantity 10 mg
Solubility Information DMSO : 62.5 mg/mL (151.19 mM; Need ultrasonic)
SMILES CN1C2=NC=NC(N3CCC3)=C2C(C4=C(C5=CC=C(C(F)(F)F)C=C5)N(C)N=C4)=N1
Molecular Weight (g/mol) 413.4
Formula Weight 413.4
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.