missing translation for 'onlineSavingsMsg'
Learn More

PF-04957325, MedChemExpress

Product Code. 16476508
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16476508 10 mg 10mg
16452607 50 mg 50mg
16441519 1 mg 1mg
16426766 100 mg 100mg
16409097 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16476508 Supplier Medchem Express Supplier No. HY15426

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PF-04957325 is a highly potent and selective PDE8 inhibitor, with IC50s of 0.7 nM and 0.3 nM for PDE8A and PDE8B, respectively.

PF-04957325 is a highly potent and selective PDE8 inhibitor, with IC50s of 0.7 nM and 0.3 nM for PDE8A and PDE8B, respectively.

Specifications

Chemical Name or Material PF-04957325
Physical Form Solid
CAS 1305115-80-3
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C14H15F3N8OS
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 100 mg/mL (249.76 mM)
SMILES NC1=C(N=NN2C[C@H]3CN(CC4=NC=CS4)CCO3)C2=NC(C(F)(F)F)=N1
Molecular Weight (g/mol) 400.38
Formula Weight 400.38
Percent Purity 98.89%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.