missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PF 04885614
GREENER_CHOICE

Product Code. 30001603
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 30001603

Brand: Tocris Bioscience™ 4916/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective NaV1.8 channel blocker; active in vivo

Potent NaV1.8 inhibitor (IC50 = 53 nM for human NaV1.8 channel). Exhibits selectivity for hNaV1.8 over hNaV1.6, hNaV1.7, hNaV1.1, hNaV1.2 and hNaV1.5 (IC50 values are 4.2, 7.0, 11, 16 and 27 μM respectively). Orally bioavailable.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1480833-70-2
Molecular Formula C13H14F3N3O

Specifications

Quantity 10 mg
Formula Weight 285.26
Purity >98%
Molecular Weight (g/mol) 285.27
Chemical Name or Material PF 04885614
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.