missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ PF 04628935

Product Code. 18788228
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18788228 5 mg 5mg
18706589 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18788228 Supplier R&D Systems™ Supplier No. 6347/5
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent ghrelin receptor inverse agonist

PF 04628935 is a potent ghrelin receptor inverse agonist (IC50 = 4.6 nM). Orally bioavailable and exhibits reasonable brain penetration.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-[2-[[2-Chloro-4-(2H-1,2,3-triazol-2-yl)phenyl]methyl]-2,7-diazaspiro[3.5]non-7-yl]-2-(2-methylimidazo[2,1-b]thiazol-6-yl)ethanone
CAS 1383719-97-8
Assay Percent Range >98%
Quantity 5 mg
Product Line Tocris
Molecular Formula C24H26ClN7OS
Formula Weight Observed MW: 496.03
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.