missing translation for 'onlineSavingsMsg'
Learn More

PF-04620110, MedChemExpress

Product Code. 16498187
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498187 5 mg 5mg
16445478 10 mg 10mg
16436066 100 mg 100mg
16411657 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16498187 Supplier Medchem Express Supplier No. HY13009
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress PF-04620110 is a potent, selective and orally bioavailable diglyceride acyltransferase-1 (DGAT-1) inhibitor with an IC50 of 19 nM.

PF-04620110 is a potent, selective and orally bioavailable diglyceride acyltransferase-1 (DGAT-1) inhibitor with an IC50 of 19 nM.

Specifications

Chemical Name or Material PF-04620110
CAS 1109276-89-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H24N4O4
Quantity 5 mg
Solubility Information DMSO : 12.5 mg/mL (31.53 mM; Need ultrasonic)
SMILES O=C(C[C@@H]1CC[C@H](CC1)C2=CC=C(C=C2)N3CCOC4=NC=NC(N)=C4C3=O)O
Molecular Weight (g/mol) 396.44
Formula Weight 396.44
Percent Purity 99.3%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.