missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PF 04620110

Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
PF 04620110 is a potent and selective DGAT-1 inhibitor (IC50 = 19 nM). Exhibits >1000-fold selectivity for DGAT-1 over DGAT-2, and no significant activity against a range of other enzymes, ion channels and receptors. Reduces plasma triglyceride levels in a rat lipid challenge model. Orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | trans-4-[4-(4-Amino-7,8-dihydro-5-oxopyrimido[5,4-f][1,4]oxazepin-6(5H)-yl)phenyl]cyclohexaneacetic acid |
| CAS | 1109276-89-2 |
| Assay Percent Range | >98% |
| Quantity | 5 mg |
| Molecular Formula | C21H24N4O4 |
| Formula Weight | Observed MW: 396.44 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction