missing translation for 'onlineSavingsMsg'
Learn More

PF 04531083, MedChemExpress

Product Code. 16486737
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16486737 5 mg 5mg
16464028 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16486737 Supplier Medchem Express Supplier No. HY105283
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress PF 04531083 is a selective NaV1.8 blocker, and used for the research of neuropathic/inflammatory pain.

PF 04531083 is a selective NaV1.8 blocker, and used for the research of neuropathic/inflammatory pain.

Specifications

Chemical Name or Material PF 04531083
CAS 1079400-07-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C17H16ClN5O2
Quantity 5 mg
Solubility Information DMSO : 125 mg/mL (349.37 mM; Need ultrasonic)
SMILES ClC1=CC=C(OC)C=C1C2=C(N=C(NC(C3=CC=NN3C)=O)C=C2)N
Molecular Weight (g/mol) 357.79
Formula Weight 357.79
Percent Purity 98.17%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.