missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PF 04479745

Product Code. 18005722
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 18005722

Brand: Tocris Bioscience™ 5755/1

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Potent and selective 5-HT2C agonist

PF 04479745 is a potent and selective 5-HT2C agonist (EC50 = 10 nM). Exhibits > 100-fold selectivity over a panel of 90 receptors, enzymes and ion channels, except for 5-HT2A (IC50 = 360 nM antagonist), 5-HT2B (EC50 = 67 nM antagonist), 5-HT1A and 5-HT6 (functional activity not tested).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (9S)-6,7,8,9-Tetrahydro-N,9-dimethyl-2-(phenylmethyl)-5H-pyrimido[4,5-d]azepin-4-amine
CAS 1065110-43-1
Quantity 1 mg
Target 5-HT2C Receptor Agonists
Molecular Formula C17H22N4
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.