missing translation for 'onlineSavingsMsg'
Learn More

PF-04447943, MedChemExpress

Product Code. 16496518
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16496518 10 mg 10mg
16459107 5 mg 5mg
16452617 50 mg 50mg
16433476 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16496518 Supplier Medchem Express Supplier No. HY15441

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PF-04447943 is a potent inhibitor of human recombinant PDE9A (IC50=12 nM) with >78-fold selectivity, respectively, over other PDE family members (IC50>1000 nM).

PF-04447943 is a potent inhibitor of human recombinant PDE9A (IC50=12 nM) with >78-fold selectivity, respectively, over other PDE family members (IC50>1000 nM).

Specifications

Chemical Name or Material PF-04447943
Physical Form Solid
CAS 1082744-20-4
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C20H25N7O2
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : ≥ 54.6 mg/mL (138.07 mM)
SMILES O=C1C2=C(N(C3CCOCC3)N=C2)N=C([C@@H]4CN(CC5=NC=CC=N5)C[C@H]4C)N1
Molecular Weight (g/mol) 395.46
Formula Weight 395.46
Percent Purity 98.74%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.