missing translation for 'onlineSavingsMsg'
Learn More

PF-04447943, MedChemExpress

Product Code. 16452617
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16452617

Brand: Medchem Express HY15441

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PF-04447943 is a potent inhibitor of human recombinant PDE9A (IC50=12 nM) with >78-fold selectivity, respectively, over other PDE family members (IC50>1000 nM).

PF-04447943 is a potent inhibitor of human recombinant PDE9A (IC50=12 nM) with >78-fold selectivity, respectively, over other PDE family members (IC50>1000 nM).

Specifications

Chemical Name or Material PF-04447943
Physical Form Solid
CAS 1082744-20-4
Color Light Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C20H25N7O2
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : ≥ 54.6 mg/mL (138.07 mM)
SMILES O=C1C2=C(N(C3CCOCC3)N=C2)N=C([C@@H]4CN(CC5=NC=CC=N5)C[C@H]4C)N1
Molecular Weight (g/mol) 395.46
Formula Weight 395.46
Percent Purity 98.74%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.