missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PF 04418948
GREENER_CHOICE

Product Code. 30001597
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001597 50 mg 50mg
30001577 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001597 Supplier Tocris Bioscience™ Supplier No. 4818/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective EP2 receptor antagonist

Potent EP2 receptor antagonist (IC50 = 16 nM for human EP2 receptors). Displays over 2000-fold selectivity for EP2 receptors over EP1, EP3, EP4, DP1 amd CRTH2 receptors; exhibits <30% binding at a diverse panel of GPCRs and ion channels at a concentration of 10 μM. Inhibits PGE2-induced increases in intracellular cAMP; reverses PGE2-invoked relaxation of mouse trachea (IC50 = 2.7 nM).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1078166-57-0
Molecular Formula C23H20FNO5

Specifications

Quantity 50 mg
Formula Weight 409.41
Purity 98%
Molecular Weight (g/mol) 409.413
Chemical Name or Material PF 04418948
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.