missing translation for 'onlineSavingsMsg'
Learn More

PF-04217903 methanesulfonate, MedChemExpress

Product Code. 16497667
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16497667 5 mg 5mg
16494918 10 mg 10mg
16481167 50 mg 50mg
16405706 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16497667 Supplier Medchem Express Supplier No. HY12017A
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress PF-04217903 methanesulfonate is a potent ATP-competitive c-Met kinase inhibitor with Ki of 4.8 nM for human c-Met. PF-04217903 methanesulfonate shows more than 1,000-fold selectivity relative to 208 kinases. Antiangiogenic properties.

PF-04217903 methanesulfonate is a potent ATP-competitive c-Met kinase inhibitor with Ki of 4.8 nM for human c-Met. PF-04217903 methanesulfonate shows more than 1,000-fold selectivity relative to 208 kinases. Antiangiogenic properties.

Specifications

Chemical Name or Material PF-04217903 methanesulfonate
CAS 956906-93-7
Color White
Purity Grade Notes Research
Molecular Formula C20H20N8O4S
Quantity 5 mg
Solubility Information DMSO : 50 mg/mL (106.73 mM; Need ultrasonic)
SMILES OCCN1N=CC(C2=NC3=C(N=C2)N=NN3CC4=CC5=CC=CN=C5C=C4)=C1.CS(=O)(O)=O
Molecular Weight (g/mol) 468.49
Formula Weight 468.49
Percent Purity 99.87%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.