missing translation for 'onlineSavingsMsg'
Learn More

PF-03814735, MedChemExpress

Product Code. 16426108
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16426108 10 mg 10mg
16408707 5 mg 5mg
16402247 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16426108 Supplier Medchem Express Supplier No. HY14574
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress PF-03814735 is a potent, orally available, ATP-competitive and reversible aurora A and aurora B inhibitor with IC50s of 0.8 and 0.5 nM, respectively.

PF-03814735 is a potent, orally available, ATP-competitive and reversible aurora A and aurora B inhibitor with IC50s of 0.8 and 0.5 nM, respectively.

Specifications

Chemical Name or Material PF-03814735
CAS 942487-16-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C23H25F3N6O2
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (210.76 mM)
SMILES O=C(CNC(C)=O)N1[C@@H]2C3=C(C=C(NC4=NC(NC5CCC5)=C(C(F)(F)F)C=N4)C=C3)[C@H]1CC2
Molecular Weight (g/mol) 474.48
Formula Weight 474.48
Percent Purity 99.03%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.