missing translation for 'onlineSavingsMsg'
Learn More

PF-03814735, MedChemExpress

Product Code. 16402247
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16402247

Brand: Medchem Express HY14574

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PF-03814735 is a potent, orally available, ATP-competitive and reversible aurora A and aurora B inhibitor with IC50s of 0.8 and 0.5 nM, respectively.

PF-03814735 is a potent, orally available, ATP-competitive and reversible aurora A and aurora B inhibitor with IC50s of 0.8 and 0.5 nM, respectively.

Specifications

Chemical Name or Material PF-03814735
CAS 942487-16-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C23H25F3N6O2
Quantity 50 mg
Solubility Information DMSO : ≥ 100 mg/mL (210.76 mM)
SMILES O=C(CNC(C)=O)N1[C@@H]2C3=C(C=C(NC4=NC(NC5CCC5)=C(C(F)(F)F)C=N4)C=C3)[C@H]1CC2
Molecular Weight (g/mol) 474.48
Formula Weight 474.48
Percent Purity 99.03%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.