missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pexmetinib, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Description
Pexmetinib is a potent Tie-2 and p38 MAPK dual inhibitor, with IC50s of 1 nM, 35 nM and 26 nM for Tie-2, p38α and p38β, respectively, and can be used in the research of acute myeloid leukemia.
Specifications
Specifications
| Chemical Name or Material | Pexmetinib |
| CAS | 945614-12-0 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C31H33FN6O3 |
| Quantity | 100 mg |
| Synonym | ARRY-614 |
| Solubility Information | DMSO : ≥ 125 mg/mL (224.57 mM) |
| SMILES | O=C(NCC1=CC(F)=CC=C1OC2=CC3=C(N(CCO)N=C3)C=C2)NC4=CC(C(C)(C)C)=NN4C5=CC=C(C)C=C5 |
| Molecular Weight (g/mol) | 556.63 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction