missing translation for 'onlineSavingsMsg'
Learn More

Pexmetinib, MedChemExpress

Product Code. 16497176
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16497176

Brand: Medchem Express HY16782

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pexmetinib is a potent Tie-2 and p38 MAPK dual inhibitor, with IC50s of 1 nM, 35 nM and 26 nM for Tie-2, p38α and p38β, respectively, and can be used in the research of acute myeloid leukemia.

Pexmetinib is a potent Tie-2 and p38 MAPK dual inhibitor, with IC50s of 1 nM, 35 nM and 26 nM for Tie-2, p38α and p38β, respectively, and can be used in the research of acute myeloid leukemia.

Specifications

Chemical Name or Material Pexmetinib
CAS 945614-12-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C31H33FN6O3
Quantity 100 mg
Synonym ARRY-614
Solubility Information DMSO : ≥ 125 mg/mL (224.57 mM)
SMILES O=C(NCC1=CC(F)=CC=C1OC2=CC3=C(N(CCO)N=C3)C=C2)NC4=CC(C(C)(C)C)=NN4C5=CC=C(C)C=C5
Molecular Weight (g/mol) 556.63
Formula Weight 556.63
Percent Purity 98.3%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.