missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PEX7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA536604
This item is not returnable.
View return policy
Description
This antibody detects endogenous protein at a molecular weight of 40 kDa. Purity is >95% by SDS-PAGE.
This gene encodes the cytosolic receptor for the set of peroxisomal matrix enzymes targeted to the organelle by the peroxisome targeting signal 2 (PTS2). Defects in this gene cause peroxisome biogenesis disorders (PBDs), which are characterized by multiple defects in peroxisome function. There are at least 14 complementation groups for PBDs, with more than one phenotype being observed in cases falling into particular complementation groups. Although the clinical features of PBD patients vary, cells from all PBD patients exhibit a defect in the import of one or more classes of peroxisomal matrix proteins into the organelle. Defects in this gene have been associated with PBD complementation group 11 (PBD-CG11) disorders, rhizomelic chondrodysplasia punctata type 1 (RCDP1), and Refsum disease (RD).
Specifications
| PEX7 | |
| Polyclonal | |
| Unconjugated | |
| PEX7 | |
| MmPEX7; PBD9B; PCDP1; Peroxin 7; Peroxin-7; peroxisomal biogenesis factor 7; peroxisomal PTS2 receptor; peroxisomal targeting signal 2 receptor; peroxisome biogenesis factor 7; peroxisome targeting signal 2 receptor; Pex7; PEX7 protein; PTS2 receptor; PTS2R; RCDP1; RD | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 18634, 308718, 5191 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide; pH 7.2 | |
| O00628, P97865 | |
| PEX7 | |
| Synthetic peptide corresponding to amino acids 211-260 of human Peroxin 7. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction