missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PEX6 Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA537881
This item is not returnable.
View return policy
Description
This antibody is predicted to react with human, mouse, pig, dog and cow based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.
PEX6 encodes a member of the AAA (ATPases associated with diverse cellular activities) family of ATPases. This member is a predominantly cytoplasmic protein, which plays a direct role in peroxisomal protein import and is required for PTS1 (peroxisomal targeting signal 1, a C-terminal tripeptide of the sequence ser-lys-leu) receptor activity. Mutations in this gene cause peroxisome biogenesis disorders of complementation group 4 and complementation group 6.
Specifications
| PEX6 | |
| Polyclonal | |
| Unconjugated | |
| Pex6 | |
| AI132582; D130055I09Rik; HMLR2; mKIAA4177; PAF 2; Paf2; Paf-2; PBD4A; PDB4B; Peroxin 6; Peroxin-6; peroxisomal AAA-type ATPase 1; peroxisomal biogenesis factor 6; Peroxisomal-type ATPase 1; peroxisome assembly factor 2; peroxisome biogenesis factor 6; PEX6; PXAAA1; RP1-125M3.3 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 117265 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P54777 | |
| Pex6 | |
| Peptide with sequence C-EQELLRYKRIQRK, from the C-terminus of PEX6 (aa 962-975). | |
| 100 μg | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction