missing translation for 'onlineSavingsMsg'
Learn More

PEX6 Antibody (S233-8), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-42199-0.025mg

Product Code. 18668237

  • £134.00 / 0.03mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PEX6 Monoclonal antibody specifically detects PEX6 in Human, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PEX6
Monoclonal
1 mg/mL
Western Blot 1:1000, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation 1:10-1:500
Q13608
Mouse
Protein G purified
RUO
Primary
Human, Rat
Purified
Western Blot, Immunofluorescence, Immunoprecipitation
S233-8
Unconjugated
PBS (pH 7.4), 50% Glycerol
PAF2, PAF-2peroxisome assembly factor 2, Peroxin-6, peroxisomal biogenesis factor 6peroxin-6, Peroxisomal-type ATPase 1, peroxisome biogenesis factor 6, PXAAA1peroxisomal AAA-type ATPase 1
Fusion protein amino acids 2-294 (N-terminus) of human PEX6. Rat: 82% identity (243/293 amino acids identical). Mouse: 82% identity (244/294 amino acids identical). <50% identity with other PEX proteins.
0.025 mg
Signal Transduction
5190
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.