missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEX6 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93534-0.1ml
This item is not returnable.
View return policy
Description
PEX6 Polyclonal antibody specifically detects PEX6 in Human, Mouse samples. It is validated for Western Blot
Specifications
| PEX6 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| PAF2, PAF-2peroxisome assembly factor 2, Peroxin-6, peroxisomal biogenesis factor 6peroxin-6, Peroxisomal-type ATPase 1, peroxisome biogenesis factor 6, PXAAA1peroxisomal AAA-type ATPase 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 741-980 of human PEX6 (NP_000278.3). LLHGPPGTGKTLLAKAVATECSLTFLSVKGPELINMYVGQSEENVREVFARARAAAPCIIFFDELDSLAPSRGRSGDSGGVMDRVVSQLLAELDGLHSTQDVFVIGATNRPDLLDPALLRPGRFDKLVFVGANEDRASQLRVLSAITRKFKLEPSVSLVNVLDCCPPQLTGADLYSLCSDAMTAALKRRVHDLEEGLEPGSSALMLTMEDLLQAAARLQPSVSEQELLRYKRIQRKFAAC | |
| 0.1 mL | |
| Signal Transduction | |
| 5190 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction