missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PEX5L Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578550
This item is not returnable.
View return policy
Description
Positive Control: mouse brain, rat brain Predicted Reactivity: Human (93%), Rhesus Monkey (94%), Bovine (85%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Pex5l is an accessory subunit of hyperpolarization-activated cyclic nucleotide-gated (HCN) channels, regulating their cell-surface expression and cyclic nucleotide dependence. Research studies have shown that Pex5l is expressed in the brain, testis and pancreas. Diseases associated with PEX5L include Rhizomelic Chondrodysplasia Punctata, Type 5 and Rhizomelic Chondrodysplasia Punctata, Type 2.
Specifications
| PEX5L | |
| Polyclonal | |
| Unconjugated | |
| PEX5L | |
| 1700016J08Rik; HCN channel auxiliary subunit; peroxin 2; peroxin-5-related protein; peroxisomal biogenesis factor 5 like; peroxisomal biogenesis factor 5-like; peroxisomal targeting signal 1 receptor-like; peroxisome biogenesis factor 5-like; Pex2; PEX2-related protein; Pex5l; PEX5-like protein; Pex5p-related protein; Pex5r; PEX5-related protein; PEX5RP; PXR2; PXR2B; Tetratricopeptide repeat-containing Rab8b-interacting protein; TPR-containing Rab8b-interacting protein; Trip8b; Trjip8b | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 286937, 58869 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.49 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q8C437, Q925N3 | |
| PEX5L | |
| Recombinant protein encompassing a sequence within the center region of mouse PEX5L. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction