missing translation for 'onlineSavingsMsg'
Learn More

PEX5 Antibody (OTI6E9) [HRP], Novus Biologicals™

Product Code. 30689078 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30689078 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30689078 Supplier Novus Biologicals Supplier No. NBP344637H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PEX5 Monoclonal antibody specifically detects PEX5 in Canine,Human,Monkey,Mouse,Rat samples. It is validated for Flow Cytometry,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen PEX5
Applications Flow Cytometry, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone OTI6E9
Conjugate HRP
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation PBS
Gene Alias FLJ50634, FLJ50721, Peroxin-5, peroxisomal biogenesis factor 5, Peroxisomal C-terminal targeting signal import receptor, peroxisomal targeting signal 1 receptor, peroxisomal targeting signal import receptor, peroxisomal targeting signal receptor 1, Peroxisome receptor 1peroxin-5, PTS1 receptor, PTS1-BP, PTS1RFLJ51948, PXR1peroxisomal targeting signal 1 (SKL type) receptor
Host Species Mouse
Immunogen Full length human recombinant protein of human PEX5 (NP_000310) produced in HEK293T cell.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5830
Target Species Canine, Human, Monkey, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.