Learn More
PEX3 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30424-1-AP-20UL
This item is not returnable.
View return policy
Description
The PEX3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PEX3. This antibody recognizes human, rat antigen. The PEX3 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| PEX3 | |
| 450 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Rat | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| DKFZp686N14184, FLJ13531, Peroxin 3, PEX3, TRG18 | |
| 20 μL | |
| 8504 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |