missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PEX3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542814
This item is not returnable.
View return policy
Description
Peptide sequence: KYGQKKIREI QEREAAEYIA QARRQYHFES NQRTCNMTVL SMLPTLREAL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
Peroxin 3, also known as PEX3 (peroxisomal biogenesis factor 3) or TRG18, is a 373 amino acid multi-pass membrane protein that localizes to peroxisomes and belongs to the Peroxin family. Expressed ubiquitously, Peroxin 3 interacts with Peroxin 19 and is involved in peroxisome biosynthesis and membrane vesicle assembly, as well as in the maintenance of peroxisomal integrity. Additionally, Peroxin 3 acts as a docking factor for Peroxin 19 and is required for the import of peroxisomal proteins. Defects in the gene encoding Peroxin 3 are the cause of peroxisome biogenesis disorder complementation group 12 (PBD-CG12) and Zellweger syndrome (ZwS), both of which arise from a failure of peroxisomal protein import.
Specifications
| PEX3 | |
| Polyclonal | |
| Unconjugated | |
| PEX3 | |
| 1700014F15Rik; 2810027F19Rik; 2900010N04Rik; PBD10A; Peroxin3; peroxin-3; peroxisomal assembly protein 3; peroxisomal assembly protein PEX3; Peroxisomal biogenesis factor 3; pex3; transformation-related protein 18; TRG18; zgc:56313 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 8504 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P56589 | |
| PEX3 | |
| Synthetic peptide directed towards the N-terminal of human PEX3 (aa 31-80). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction