missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEX26 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-32743
This item is not returnable.
View return policy
Description
PEX26 Polyclonal specifically detects PEX26 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| PEX26 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris-Glycine (pH7), 10% Glycerol with 0.01% Thimerosal | |
| PEX26 | |
| Full length human PEX26 Recombinant protein. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| FLJ20695, peroxin-26, peroxisomal biogenesis factor 26, peroxisome assembly protein 26, peroxisome biogenesis disorder, complementation group 8, peroxisome biogenesis disorder, complementation group A, peroxisome biogenesis factor 26, PEX26M1T, Pex26pM1T | |
| Rabbit | |
| 34 kDa | |
| 100 μL | |
| Signal Transduction | |
| 55670 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction