missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEX26 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-52048
This item is not returnable.
View return policy
Description
PEX26 Polyclonal specifically detects PEX26 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| PEX26 | |
| Polyclonal | |
| Western Blot 0.5 - 1.5 ug/ml, Peptide ELISA Detection limit 1:64000 | |
| NP_060399.1 | |
| PEX26 | |
| Peptide with sequence C-QKPNLEGSVSHK corresponding to internal region according to NP_060399.1. | |
| 0.1 mg | |
| Signal Transduction | |
| 55670 | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| FLJ20695, peroxin-26, peroxisomal biogenesis factor 26, peroxisome assembly protein 26, peroxisome biogenesis disorder, complementation group 8, peroxisome biogenesis disorder, complementation group A, peroxisome biogenesis factor 26, PEX26M1T, Pex26pM1T | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Reported variants represent identical protein: NP_060399.1, NP_001121121.1 | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction