missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PEX2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552832
This item is not returnable.
View return policy
Description
Immunogen sequence: NAKSANRVLR ISQLDALELN KALEQLVWSQ FTQCFHGFKP GLLARFEPEV KACLWVFLWR FTIYSKNATV GQSVLNIKYK NDFSPNLRYQ PPSKNQKIWY AVCTIGGRWL EERCYDLFRN HHLASFGKVK Q Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 90%.
This gene encodes an integral peroxisomal membrane protein required for peroxisome biogenesis. The protein is thought to be involved in peroxisomal matrix protein import. Mutations in this gene result in one form of Zellweger syndrome and infantile Refsum disease. Alternative splicing results in multiple transcript variants encoding the same protein.
Specifications
| PEX2 | |
| Polyclonal | |
| Unconjugated | |
| PEX2 | |
| 35 kDa peroxisomal membrane protein; D3Ertd138e; Paf1; PAF-1; PBD5A; PBD5B; peroxin 2; Peroxin-2; peroxisomal biogenesis factor 2; Peroxisomal membrane protein 3; peroxisomal membrane protein 3, 35 kDa; peroxisomal membrane protein 3, 35kDa; peroxisome assembly factor 1; peroxisome assembly factor-1; peroxisome biogenesis factor 2; Pex2; PMP3; Pmp35; Pxmp3; RING finger protein 72; RNF72; Zellweger syndrome homolog; ZWS3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5828 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P28328 | |
| PEX2 | |
| Recombinant protein corresponding to Human PEX2. Recombinant protein control fragment (Product #RP-90398). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction