missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PEX2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA519321
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.
This gene encodes an integral peroxisomal membrane protein required for peroxisome biogenesis. The protein is thought to be involved in peroxisomal matrix protein import. Mutations in this gene result in one form of Zellweger syndrome and infantile Refsum disease. Alternative splicing results in multiple transcript variants encoding the same protein.
Specifications
| PEX2 | |
| Polyclonal | |
| Unconjugated | |
| PEX2 | |
| 35 kDa peroxisomal membrane protein; D3Ertd138e; Paf1; PAF-1; PBD5A; PBD5B; peroxin 2; Peroxin-2; peroxisomal biogenesis factor 2; Peroxisomal membrane protein 3; peroxisomal membrane protein 3, 35 kDa; peroxisomal membrane protein 3, 35kDa; peroxisome assembly factor 1; peroxisome assembly factor-1; peroxisome biogenesis factor 2; Pex2; PMP3; Pmp35; Pxmp3; RING finger protein 72; RNF72; Zellweger syndrome homolog; ZWS3 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 5828 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P28328 | |
| PEX2 | |
| Synthetic peptide sequence (NATVGQSVLNIKYKN) corresponding to the internal amino acids of PXMP3 (aa 73-88). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction