missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PEX19 Monoclonal Antibody (GT554)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA517266
This item is not returnable.
View return policy
Description
Recommended positive controls: Jurkat, Raji, K562, THP-1, HL-60, NCI-H929, mouse liver. Predicted reactivity: Mouse (91%), Rat (93%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene is necessary for early peroxisomal biogenesis. It acts both as a cytosolic chaperone and as an import receptor for peroxisomal membrane proteins (PMPs). Peroxins (PEXs) are proteins that are essential for the assembly of functional peroxisomes. The peroxisome biogenesis disorders (PBDs) are a group of genetically heterogeneous autosomal recessive, lethal diseases characterized by multiple defects in peroxisome function. The peroxisomal biogenesis disorders are a heterogeneous group with at least 14 complementation groups and with more than 1 phenotype being observed in cases falling into particular complementation groups. Although the clinical features of PBD patients vary, cells from all PBD patients exhibit a defect in the import of one or more classes of peroxisomal matrix proteins into the organelle. Defects in this gene are a cause Zellweger syndrome (ZWS).
Specifications
| PEX19 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P40855, Q8VCI5 | |
| PEX19 | |
| Recombinant protein encompassing a sequence within the center region of human PEX19. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| GT554 | |
| Unconjugated | |
| PEX19 | |
| 33 kDa housekeeping protein; D1S2223E; HK33; housekeeping gene, 33kD; OK/SW-cl.22; PBD12A; peroxin-19; Peroxisomal biogenesis factor 19; peroxisomal farnesylated protein; peroxisome biogenesis factor 19; Pex19; PMP1; PMPI; PxF; PXMP1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 19298, 5824 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction