missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEX16 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68261-1-IG-20UL
This item is not returnable.
View return policy
Description
The PEX16 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PEX16. This antibody recognizes human, rat, mouse antigen. The PEX16 antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| PEX16 | |
| Monoclonal | |
| 500 μg/mL | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 9409 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG1 |
| Western Blot, ELISA | |
| 2D7G4 | |
| Unconjugated | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| 38 kDa | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction