missing translation for 'onlineSavingsMsg'
Learn More

PEX14 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16887073 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16887073 150 μL 150µL
16897073 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16887073 Supplier Proteintech Supplier No. 105941AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody can be used to detect endogenous PEX14 with an apparent molecular weight of 57 kDa and recognize peroxisomal structures in human, monkey and mouse cells.

PEX14 belongs to the peroxin 14 family. It is a component of the peroxisomal translocation machinery with PEX13 and PEX17 and interacts with both the PTS1 and PTS2 receptors. PEX14 binds directly to PEX17. Defects in PEX14 are the cause of peroxisome biogenesis disorder complementation group K (PBD-CGK) [MIM:601791] and Zellweger syndrome (ZWS) [MIM:214100].

Specifications

Antigen PEX14
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene PEX14
Gene Accession No. O75381, Q642G4, Q9R0A0
Gene Alias NAPP2, Peroxin 14, PEX14, Pex14p, PTS1 receptor docking protein
Gene Symbols PEX14
Host Species Rabbit
Immunogen PEX14 Fusion Protein Ag0932
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5195, 56273, 64460
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.