missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PEX12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568265
This item is not returnable.
View return policy
Description
Predicted to react with Mouse and Rat samples.
Peroxins (PEXs) are proteins that are essential for the assembly of functional peroxisomes. The peroxisome biogenesis disorders (PBDs) are a group of genetically heterogeneous autosomal recessive, lethal diseases characterized by multiple defects in peroxisome function. The peroxisomal biogenesis disorders are a heterogeneous group with at least 14 complementation groups and with more than 1 phenotype being observed in cases falling into particular complementation groups. Although the clinical features of PBD patients vary, cells from all PBD patients exhibit a defect in the import of one or more classes of peroxisomal matrix proteins into the organelle.
Specifications
| PEX12 | |
| Polyclonal | |
| Unconjugated | |
| PEX12 | |
| AI451906; PAF3; PAF-3; PBD3A; peroxin 12; peroxin-12; peroxisomal biogenesis factor 12; peroxisome assembly factor 3; Peroxisome assembly protein 12; Pex12 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 103737, 116718, 5193 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O00623, O88177, Q8VC48 | |
| PEX12 | |
| Synthesized peptide derived from human PEX12. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction