missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PEX11G Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA575897
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Peroxisomes are single-membrane bound organelles present in virtually all eukaryotic cells. They are involved in numerous catabolic and anabolic pathways, including beta-oxidation of very long chain fatty acids, metabolism of hydrogen peroxide, plasmalogen biosynthesis and bile acid synthesis. The Peroxin family, which includes more than 20 members, is required for peroxisome biogenesis. The Peroxin 11 peroxisomal membrane proteins are the only factors known to promote peroxisome division in multiple species. Peroxin 11 proteins have a direct role in peroxisomal fatty acid oxidation and only affect peroxisome abundance indirectly. Peroxin 11gamma (peroxisomal biogenesis factor 11 gamma), also known as PEX11G or Peroxin-11C, is a 241 amino acid multi-pass membrane protein that belongs to the peroxin 11 family. Peroxin 11gamma exist as two isoforms due to alternative splicing events.
Specifications
| PEX11G | |
| Polyclonal | |
| Unconjugated | |
| PEX11G | |
| 1810022F11Rik; 1810049N02Rik; peroxin Pex11p gamma; peroxin-11C; peroxisomal biogenesis factor 11 gamma; peroxisomal biogenesis factor 11c; peroxisomal biogenesis factor 11G; peroxisomal membrane protein 11C; Pex11c; Pex11g; Pex11gamma; PEX11-gamma; Pex11pgamma; protein PEX11 homolog gamma | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 288369, 69129, 92960 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6P6M5, Q96HA9 | |
| PEX11G | |
| Synthetic peptide corresponding to residues in Human PEX11G. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction