missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PEX11B Recombinant Rabbit Monoclonal Antibody (K01_1W63)
Description
PEX11B Recombinant Monoclonal Antibody for Western Blot
Peroxisomal membrane protein 11B is a protein that in humans is encoded by the PEX11B gene. Involved in peroxisomal proliferation. May regulate peroxisomes division by recruiting the dynamin-related GTPase DNM1L to the peroxisomal membrane.
Specifications
Specifications
| Antigen | PEX11B |
| Applications | Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | K01_1W63 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM tris glycine with 150mM NaCl, 0.05% BSA, 40% glycerol and 0.01% sodium azide; pH 7.4 |
| Gene | PEX11B |
| Gene Accession No. | O96011, Q9Z210 |
| Gene Alias | Peroxin 11 beta; Peroxin 11B; Peroxin-11 beta; Peroxin11B; peroxin-11B; peroxisomal biogenesis factor 11 beta; peroxisomal biogenesis factor 11B; Peroxisomal membrane protein 11B; PEX11 beta; PEX-11 beta; PEX11B; PEX11beta; PEX11-beta; Pex11pbeta; PEX14B; Protein PEX11 homolog beta |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?