missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEX11B Antibody (S09-7B4), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19737-50ul
This item is not returnable.
View return policy
Description
PEX11B Monoclonal antibody specifically detects PEX11B in Human, Mouse samples. It is validated for Western Blot
Specifications
| PEX11B | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 8799 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| S09-7B4 | |
| Unconjugated | |
| peroxin-11B, peroxisomal biogenesis factor 11 beta, Peroxisomal biogenesis factor 11Bperoxisomal membrane protein 11B, PEX11-beta, Protein PEX11 homolog beta | |
| A synthetic peptide of human PEX11B (Uniprot # O96011) | |
| 50 μg | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction