missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEX11B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35819-100ul
This item is not returnable.
View return policy
Description
PEX11B Polyclonal antibody specifically detects PEX11B in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| PEX11B | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| peroxin-11B, peroxisomal biogenesis factor 11 beta, Peroxisomal biogenesis factor 11Bperoxisomal membrane protein 11B, PEX11-beta, Protein PEX11 homolog beta | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 100-180 of human PEX11B (NP_003837.1).,, Sequence:, DNVLWAGKSGLAPRVDQEKWAQRSFRYYLFSLIMNLSRDAYEIRLLMEQESSACSRRLKGSGGGVPGGSETGGLGGPGTPG | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 8799 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction