missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEX10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35707-20ul
This item is not returnable.
View return policy
Description
PEX10 Polyclonal antibody specifically detects PEX10 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| PEX10 | |
| Polyclonal | |
| Western Blot 1:1000 - 1:5000, ELISA | |
| NALD, peroxin 10, Peroxin-10, peroxisomal biogenesis factor 10MGC1998, Peroxisome assembly protein 10, RING finger protein 69, RNF69peroxisome biogenesis factor 10 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 10-160 of human PEX10 (NP_002608.1).,, Sequence:, EVIRAAQKDEYYRGGLRSAAGGALHSLAGARKWLEWRKEVELLSDVAYFGLTTLAGYQTLGEEYVSIIQVDPSRIHVPSSLRRGVLVTLHAVLPYLLDKALLPLEQELQADPDSGRPLQGSLGPGGRGCSGARRWMRHHTATLTEQQRRAL | |
| 20 μL | |
| Zinc Finger | |
| 5192 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction