missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ PEX10 Antibody Blocking Peptide

Product Code. 18315283 Shop All Bio Techne Products
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
This item is not returnable. View return policy

Product Code. 18315283

Brand: Novus Biologicals™ NBP318137PEP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Blocking Peptied

TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.2 mg/ml
For Use With (Application) Antibody Competition
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin
Gene ID (Entrez) 5192
Quantity 0.05 mg
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Gene Alias NALD, peroxin 10, Peroxin-10, peroxisomal biogenesis factor 10MGC1998, Peroxisome assembly protein 10, RING finger protein 69, RNF69peroxisome biogenesis factor 10
Gene Symbol PEX10
Research Category Zinc Finger
Protein PEX10
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.