missing translation for 'onlineSavingsMsg'
Learn More

PEX10 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18339075 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18339075 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18339075 Supplier Novus Biologicals Supplier No. NBP318137AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PEX10 Polyclonal antibody specifically detects PEX10 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen PEX10
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias NALD, peroxin 10, Peroxin-10, peroxisomal biogenesis factor 10MGC1998, Peroxisome assembly protein 10, RING finger protein 69, RNF69peroxisome biogenesis factor 10
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 10 amino acids near the carboxyl terminus of human PEX10. The immunogen is located within last 50 amino acids of PEX10.
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 5192
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.