missing translation for 'onlineSavingsMsg'
Learn More

PEX10 Antibody (1B8), Novus Biologicals™

Product Code. 18335919 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18335919 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18335919 Supplier Novus Biologicals Supplier No. H00005192M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PEX10 Monoclonal antibody specifically detects PEX10 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen PEX10
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 1B8
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH18198
Gene Alias NALD, peroxin 10, Peroxin-10, peroxisomal biogenesis factor 10MGC1998, Peroxisome assembly protein 10, RING finger protein 69, RNF69peroxisome biogenesis factor 10
Host Species Mouse
Immunogen PEX10 (AAH18198.1, 1 a.a. ~ 326 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MAPAAASPPEVIRAAQKDEYYRGGLRSAAGGALHSLAGARKWLEWRKEVELLSDVAYFGLTTLAGYQTLGEEYVSIIQVDPSRIHVPSSLRRGVLVTLHAVLPYLLDKALLPLEQELQADPDSGRPLQGSLGPGGRGCSGARRWMRHHTATLTEQQRRALLRAVFVLRQGLACLQRLHVAWFYIHGVFYHLAKRLTGITYLRVRSLPGEDLRARVSYRLLGVISLLHLVLSMGLQLYGFRQRQRARKEWRLHRGLSHRRASLEERAVSRNPLCTLCLEERRHPTATPCGHLFCWECITAWCSSKAECPLCREKFPPQKLIYLRHYR
Purification Method IgG purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 5192
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.