missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PEX1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30128
This item is not returnable.
View return policy
Description
PEX1 Polyclonal antibody specifically detects PEX1 in Human samples. It is validated for Immunoprecipitation
Specifications
| PEX1 | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 5189 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Immunoprecipitation | |
| Unconjugated | |
| peroxin-1, peroxisomal biogenesis factor 1, Peroxisome biogenesis disorder protein 1, peroxisome biogenesis factor 1, Zellweger syndrome, Zellweger syndrome 1, ZWS, ZWS1 | |
| Between 1100 and 1150 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction