missing translation for 'onlineSavingsMsg'
Learn More

Pevonedistat, MedChemExpress

Product Code. 16448188
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
1mg
200mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16448188 10 mg 10mg
16491336 200 mg 200mg
16481959 1 mg 1mg
16464087 50 mg 50mg
16460628 5 mg 5mg
16437936 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16448188 Supplier Medchem Express Supplier No. HY70062
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Pevonedistat (MLN4924) is a potent and selective NEDD8-activating enzyme (NAE) inhibitor with an IC50 of 4.7 nM.

Pevonedistat (MLN4924) is a potent and selective NEDD8-activating enzyme (NAE) inhibitor with an IC50 of 4.7 nM.

Specifications

Chemical Name or Material Pevonedistat
CAS 905579-51-3
Color White
Purity Grade Notes Research
Molecular Formula C21H25N5O4S
Quantity 10 mg
Synonym MLN4924
Solubility Information DMSO : 62.5 mg/mL (140.92 mM; Need ultrasonic)
SMILES O=S(OC[C@@H]1C[C@@H](N2C3=NC=NC(N[C@H]4CCC5=C4C=CC=C5)=C3C=C2)C[C@@H]1O)(N)=O
Molecular Weight (g/mol) 443.52
Formula Weight 443.52
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.